Product Details

SNP ID
rs17255807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59490441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTATTTGAGTGAATCGTTTATTG[A/G]TGAAGGTCTGGTGTATGGAAGTGGC
Phenotype
MIM: 611176 MIM: 614811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JKAMP PubMed Links

Gene Details

Gene
JKAMP
Gene Name
JNK1/MAPK8-associated membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098625.2 Intron NP_001092095.1
NM_001284201.1 Intron NP_001271130.1
NM_001284202.1 Intron NP_001271131.1
NM_001284203.1 Intron NP_001271132.1
NM_001284204.1 Intron NP_001271133.1
NM_016475.4 Intron NP_057559.2
XM_006720171.3 Intron XP_006720234.1
XM_017021366.1 Intron XP_016876855.1
Gene
L3HYPDH
Gene Name
trans-L-3-hydroxyproline dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144581.1 Intron NP_653182.1

View Full Product Details