Product Details

SNP ID
rs16995485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18199868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTATATTCTCCACCCTACACTT[C/G]CAGATATTTGTTTCTATATGGTTAT
Phenotype
MIM: 616133 MIM: 600128 MIM: 179490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC102725254 PubMed Links

Gene Details

Gene
LOC102725254
Gene Name
uncharacterized LOC102725254
There are no transcripts associated with this gene.

Gene
MPV17L2
Gene Name
MPV17 mitochondrial inner membrane protein like 2
There are no transcripts associated with this gene.

Gene
PDE4C
Gene Name
phosphodiesterase 4C
There are no transcripts associated with this gene.

Gene
RAB3A
Gene Name
RAB3A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002866.4 Intron NP_002857.1
XM_011528164.1 Intron XP_011526466.1

View Full Product Details