Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005116.5 | 4554 | UTR 3 | NP_005107.4 | ||
NM_203327.1 | 4554 | UTR 3 | NP_976072.1 | ||
XM_011529414.2 | 4554 | UTR 3 | XP_011527716.1 | ||
XM_011529415.2 | 4554 | UTR 3 | XP_011527717.1 | ||
XM_011529416.2 | 4554 | UTR 3 | XP_011527718.1 | ||
XM_011529417.2 | 4554 | UTR 3 | XP_011527719.1 | ||
XM_017028171.1 | 4554 | UTR 3 | XP_016883660.1 | ||
XM_017028172.1 | 4554 | UTR 3 | XP_016883661.1 | ||
XM_017028173.1 | 4554 | UTR 3 | XP_016883662.1 | ||
XM_017028174.1 | 4554 | UTR 3 | XP_016883663.1 | ||
XM_017028175.1 | 4554 | UTR 3 | XP_016883664.1 | ||
XM_017028176.1 | 4554 | UTR 3 | XP_016883665.1 | ||
XM_017028177.1 | 4554 | UTR 3 | XP_016883666.1 | ||
XM_017028178.1 | 4554 | UTR 3 | XP_016883667.1 | ||
XM_017028179.1 | 4554 | UTR 3 | XP_016883668.1 | ||
XM_017028180.1 | 4554 | UTR 3 | XP_016883669.1 | ||
XM_017028181.1 | 4554 | Intron | XP_016883670.1 |