Product Details
- SNP ID
-
rs6799856
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:125451536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCCTATGTAGAAGGAAGAGTTAT[C/T]GCTTGTATAAGACAGTTTTTTTATT
- Phenotype
-
MIM: 605931
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX4
PubMed Links
Gene Details
- Gene
- SNX4
- Gene Name
- sorting nexin 4
View Full Product Details