Product Details
- SNP ID
-
rs7653955
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:44620608 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGATTACATAGTCAAAATAATT[A/G]ATGGACTAGATATTGGATGTGAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YIPF7
PubMed Links
Gene Details
- Gene
- YIPF7
- Gene Name
- Yip1 domain family member 7
View Full Product Details