Product Details

SNP ID
rs7686422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:9773803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAACTCTTTGTGGGAAAGAGCC[A/T]GGGTAAGAGAGGACACCCACTACTT
Phenotype
MIM: 126453 MIM: 606142
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DRD5 PubMed Links

Gene Details

Gene
DRD5
Gene Name
dopamine receptor D5
There are no transcripts associated with this gene.

Gene
SLC2A9
Gene Name
solute carrier family 2 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001290.1 Intron NP_001001290.1
NM_020041.2 Intron NP_064425.2
XM_006713968.3 Intron XP_006714031.1
XM_006713969.2 Intron XP_006714032.1
XM_011513856.2 Intron XP_011512158.1
XM_011513857.1 Intron XP_011512159.1
XM_011513858.1 Intron XP_011512160.1
XM_011513859.2 Intron XP_011512161.1
XM_011513860.2 Intron XP_011512162.1
XM_011513861.2 Intron XP_011512163.1
XM_011513862.2 Intron XP_011512164.1
XM_011513863.2 Intron XP_011512165.1
XM_011513864.2 Intron XP_011512166.1
XM_011513865.2 Intron XP_011512167.1
XM_011513866.2 Intron XP_011512168.1
XM_011513867.2 Intron XP_011512169.1
XM_011513868.2 Intron XP_011512170.1
XM_017008457.1 Intron XP_016863946.1
XM_017008458.1 Intron XP_016863947.1
XM_017008459.1 Intron XP_016863948.1
XM_017008460.1 Intron XP_016863949.1

View Full Product Details