Product Details

SNP ID
rs13238725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:727678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCACTCCCCTGTCCCCTTGCTC[A/C]CCGCCTCCACCCTCCACGCCCCAAG
Phenotype
MIM: 614864 MIM: 176911
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAAF5 PubMed Links

Gene Details

Gene
DNAAF5
Gene Name
dynein axonemal assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017802.3 Intron NP_060272.3
Gene
PRKAR1B
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164758.1 Intron NP_001158230.1
NM_001164759.1 Intron NP_001158231.1
NM_001164760.1 Intron NP_001158232.1
NM_001164761.1 Intron NP_001158233.1
NM_001164762.1 Intron NP_001158234.1
NM_002735.2 Intron NP_002726.1
XM_011515446.2 Intron XP_011513748.1
XM_017012433.1 Intron XP_016867922.1
XM_017012434.1 Intron XP_016867923.1

View Full Product Details