Product Details

SNP ID
rs11542253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33817814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCTGATGCTCGAGCTGAAATC[A/C]CTGCAGGAGGAACCGGTGGAGGGCT
Phenotype
MIM: 612506
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE2R2 PubMed Links

Gene Details

Gene
UBE2R2
Gene Name
ubiquitin conjugating enzyme E2 R2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017811.3 651 Silent Mutation TCA,TCC S19S NP_060281.2
XM_005251496.2 651 Silent Mutation TCA,TCC S19S XP_005251553.1
XM_011517949.1 651 Silent Mutation TCA,TCC S19S XP_011516251.1
XM_017014860.1 651 Silent Mutation TCA,TCC S19S XP_016870349.1

View Full Product Details