Product Details

SNP ID
rs7178663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90353826 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGCTGAGGCTGGTTTATTGGAAA[C/T]TTTTTAAGGGAAAAAACATTATCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABARAPL3 PubMed Links

Gene Details

Gene
GABARAPL3
Gene Name
GABA type A receptor associated protein like 3 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF774
Gene Name
zinc finger protein 774
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004309.2 Intron NP_001004309.2
XM_017022127.1 Intron XP_016877616.1
XM_017022128.1 Intron XP_016877617.1
XM_017022129.1 Intron XP_016877618.1

View Full Product Details