Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001126102.2 | 796 | Silent Mutation | GCC,GCT | A245A | NP_001119574.1 |
NM_001318138.1 | 796 | Silent Mutation | GCC,GCT | A245A | NP_001305067.1 |
NM_005143.4 | 796 | Silent Mutation | GCC,GCT | A304A | NP_005134.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142317.1 | 796 | Intron | NP_001135789.1 | ||
NM_001142318.1 | 796 | Intron | NP_001135790.1 | ||
NM_001324354.1 | 796 | Intron | NP_001311283.1 | ||
NM_001324355.1 | 796 | Intron | NP_001311284.1 | ||
NM_017853.2 | 796 | Intron | NP_060323.1 | ||
XM_017023377.1 | 796 | Intron | XP_016878866.1 |