Product Details

SNP ID
rs7198286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87888557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCCATAGGGAGAGGAACTGAGG[C/T]TCCCAGCGAACTGCCAGCACCAACT
Phenotype
MIM: 114761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA5A PubMed Links

Gene Details

Gene
CA5A
Gene Name
carbonic anhydrase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001739.1 Intron NP_001730.1
XM_005256134.4 Intron XP_005256191.1
XM_011523309.2 Intron XP_011521611.1
XM_011523310.2 Intron XP_011521612.1
XM_017023646.1 Intron XP_016879135.1

View Full Product Details