Product Details

SNP ID
rs9896940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39739722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACGGGAACGCGGGTGGGGCTGGA[A/G]CCACTGGGGGAGGAGGCAGCTGGGA
Phenotype
MIM: 601522 MIM: 611802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRB7 PubMed Links

Gene Details

Gene
GRB7
Gene Name
growth factor receptor bound protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030002.2 704 Intron NP_001025173.1
NM_001242442.1 704 Intron NP_001229371.1
NM_001242443.1 704 Intron NP_001229372.1
NM_005310.3 704 Intron NP_005301.2
XM_017024536.1 704 UTR 5 XP_016880025.1
XM_017024537.1 704 Intron XP_016880026.1
XM_017024538.1 704 Intron XP_016880027.1
Gene
MIEN1
Gene Name
migration and invasion enhancer 1
There are no transcripts associated with this gene.

View Full Product Details