Product Details

SNP ID
rs8078399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40068245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTTTCATTATCTAGAGACAATG[G/T]TTCCTGTCTCACTCAGGTTGAGGGC
Phenotype
MIM: 190120
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
THRA PubMed Links

Gene Details

Gene
THRA
Gene Name
thyroid hormone receptor, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190918.1 Intron NP_001177847.1
NM_001190919.1 Intron NP_001177848.1
NM_003250.5 Intron NP_003241.2
NM_199334.3 Intron NP_955366.1

View Full Product Details