Product Details

SNP ID
rs11541535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2476583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTTGGTGGCCGCTCAGCGCCA[C/G]GATCGCCTCACAGTGGGGGTGTACG
Phenotype
MIM: 604948
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GADD45B PubMed Links

Gene Details

Gene
GADD45B
Gene Name
growth arrest and DNA damage inducible beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015675.3 333 Missense Mutation CAC,CAG H33Q NP_056490.2
XM_017026822.1 333 Missense Mutation CAC,CAG H33Q XP_016882311.1

View Full Product Details