Product Details

SNP ID
rs13433602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31115213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACCCAGTAATTCCATTTTTCA[A/G]AATTAGCCCCAAAGTAATCATCAGA
Phenotype
MIM: 606481 MIM: 610918
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5J PubMed Links

Gene Details

Gene
INPP5J
Gene Name
inositol polyphosphate-5-phosphatase J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002837.2 Intron NP_001002837.1
NM_001284285.1 Intron NP_001271214.1
NM_001284286.1 Intron NP_001271215.1
NM_001284287.1 Intron NP_001271216.1
NM_001284288.1 Intron NP_001271217.1
NM_001284289.1 Intron NP_001271218.1
XM_011530143.1 Intron XP_011528445.1
XM_017028771.1 Intron XP_016884260.1
XM_017028772.1 Intron XP_016884261.1
XM_017028773.1 Intron XP_016884262.1
Gene
SELM
Gene Name
selenoprotein M
There are no transcripts associated with this gene.

View Full Product Details