Product Details

SNP ID
hCV43984224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31758159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAAATTCTTTGAACATTGAATA[A/C]CTGTAACGAATTGTGCTGTATCTGT
Phenotype
MIM: 614191
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DEPDC5 PubMed Links

Gene Details

Gene
DEPDC5
Gene Name
DEP domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007188.2 Intron NP_001007189.1
NM_001136029.2 Intron NP_001129501.1
NM_001242896.1 Intron NP_001229825.1
NM_001242897.1 Intron NP_001229826.1
NM_014662.4 Intron NP_055477.1
Gene
PRR14L
Gene Name
proline rich 14 like
There are no transcripts associated with this gene.

View Full Product Details