Product Details

SNP ID
rs8138116
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32123032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCGCTATCCTGGGCTGCTACTCC[C/T]GGTGGGAGCAGATTGGAGCGCTATC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AP1B1P1 PubMed Links

Gene Details

Gene
AP1B1P1
Gene Name
adaptor related protein complex 1 beta 1 subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details