Product Details

SNP ID
rs12389413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53071408 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTCAACAACTCTAAGGAAAGCT[C/T]ATCCCAAACCAAAGTGCAGCCTCCT
Phenotype
MIM: 300253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR173 PubMed Links

Gene Details

Gene
GPR173
Gene Name
G protein-coupled receptor 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018969.5 Intron NP_061842.1
XM_011530798.2 Intron XP_011529100.1

View Full Product Details