Product Details

SNP ID
rs7890070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:19350162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTATTTAGGTACTGAAGTATGG[C/T]TTGTACTTATTGGGCTTTACCCTGC
Phenotype
MIM: 300820 MIM: 300502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K15 PubMed Links

Gene Details

Gene
MAP3K15
Gene Name
mitogen-activated protein kinase kinase kinase 15
There are no transcripts associated with this gene.

Gene
PDHA1
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000284.3 Intron NP_000275.1
NM_001173454.1 Intron NP_001166925.1
NM_001173455.1 Intron NP_001166926.1
NM_001173456.1 Intron NP_001166927.1
XM_017029574.1 Intron XP_016885063.1

View Full Product Details