Product Details

SNP ID
rs3097085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:7289695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAAATATAAAAAGAAAGAAAAC[A/G]GAAAAAAAAATATATTTATATAGAA
Phenotype
MIM: 400008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKY PubMed Links

Gene Details

Gene
PRKY
Gene Name
protein kinase, Y-linked, pseudogene
There are no transcripts associated with this gene.

View Full Product Details