Product Details

SNP ID
rs11546701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99885639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCAAAAGTGGATAAGAAACCA[G/T]GTCGCAAAGCATGCTCTGGAGCATT
Phenotype
MIM: 603296
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAMTOR3 PubMed Links

Gene Details

Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243736.1 389 Missense Mutation CAT,CCT H40P NP_001230665.1
NM_021970.3 389 Missense Mutation CAT,CCT H47P NP_068805.1

View Full Product Details