Product Details

SNP ID
rs7654331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77714115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAGGGAAAAAGTTACTAAAAC[A/C]CCTTGCTTGGTTTTACAGTCATGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNOT6L PubMed Links

Gene Details

Gene
CNOT6L
Gene Name
CCR4-NOT transcription complex subunit 6 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286790.1 8092 UTR 3 NP_001273719.1
NM_144571.2 8092 UTR 3 NP_653172.2
XM_011531806.2 8092 UTR 3 XP_011530108.1
XM_011531807.2 8092 UTR 3 XP_011530109.1
XM_011531808.2 8092 UTR 3 XP_011530110.1
XM_011531809.1 8092 UTR 3 XP_011530111.1
XM_011531810.1 8092 UTR 3 XP_011530112.1
XM_017007956.1 8092 UTR 3 XP_016863445.1
XM_017007957.1 8092 UTR 3 XP_016863446.1

View Full Product Details