Product Details
- SNP ID
-
rs7558887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100275971 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACATGCAGATAGGCTATTTTAAC[G/T]CAAATATATAATAAATTGTACATGT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LONRF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148310023] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LONRF2
- Gene Name
- LON peptidase N-terminal domain and ring finger 2
View Full Product Details