Product Details

SNP ID
rs11555864
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37344758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGCGGAAGACACCGGCGCGT[A/C]GTGGGCACCCTCCACCTGCTGCTGC
Phenotype
MIM: 607065
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
QPCT PubMed Links

Gene Details

Gene
QPCT
Gene Name
glutaminyl-peptide cyclotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012413.3 149 Silent Mutation GTA,GTC V9V NP_036545.1

View Full Product Details