Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007102.2 | Intron | NP_001007103.1 | |||
NM_032438.2 | Intron | NP_115814.1 | |||
XM_005267161.4 | Intron | XP_005267218.1 | |||
XM_006715576.3 | Intron | XP_006715639.1 | |||
XM_006715578.3 | Intron | XP_006715641.1 | |||
XM_011536179.2 | Intron | XP_011534481.1 | |||
XM_011536180.2 | Intron | XP_011534482.1 | |||
XM_011536181.2 | Intron | XP_011534483.1 | |||
XM_011536183.2 | Intron | XP_011534485.1 | |||
XM_011536184.2 | Intron | XP_011534486.1 | |||
XM_017011355.1 | Intron | XP_016866844.1 | |||
XM_017011356.1 | Intron | XP_016866845.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001017373.3 | Intron | NP_001017373.2 | |||
NM_001258275.2 | Intron | NP_001245204.1 | |||
NM_001277185.1 | Intron | NP_001264114.1 | |||
XM_017010305.1 | Intron | XP_016865794.1 | |||
XM_017010306.1 | Intron | XP_016865795.1 | |||
XM_017010307.1 | Intron | XP_016865796.1 | |||
XM_017010308.1 | Intron | XP_016865797.1 | |||
XM_017010309.1 | Intron | XP_016865798.1 | |||
XM_017010310.1 | Intron | XP_016865799.1 |