Product Details
- SNP ID
-
rs5029929
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:137869500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGCCCAATTCCACCTGTAGAGGA[C/T]GTCTGACATCAAAAGAGAGCACCAT
- Phenotype
-
MIM: 191163
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100130476
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5029930] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100130476
- Gene Name
- uncharacterized LOC100130476
There are no transcripts associated with this gene.
- Gene
- LOC107986649
- Gene Name
- uncharacterized LOC107986649
There are no transcripts associated with this gene.
- Gene
- TNFAIP3
- Gene Name
- TNF alpha induced protein 3
View Full Product Details