Product Details

SNP ID
rs1254974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52744496 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTAGGTATCTGCTGTCAGTTGAT[C/T]AGAGTTGGCATGAGAAACAAAAAAA
Phenotype
MIM: 615814
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STYX PubMed Links

Gene Details

Gene
STYX
Gene Name
serine/threonine/tyrosine interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130701.1 Intron NP_001124173.1
NM_145251.3 Intron NP_660294.1
XM_006720242.3 Intron XP_006720305.1
XM_011537108.2 Intron XP_011535410.1
XM_011537109.2 Intron XP_011535411.1

View Full Product Details