Product Details

SNP ID
hCV44825826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150446929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATTCATCTCGGTCTGCCTTTA[A/C]CTTCATCCCACCAGTCACACGGCAG
Phenotype
MIM: 130620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPS14 PubMed Links

Gene Details

Gene
RPS14
Gene Name
ribosomal protein S14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025070.1 457 Missense Mutation GTA,TTA V62L NP_001020241.1
NM_001025071.1 457 Missense Mutation GTA,TTA V62L NP_001020242.1
NM_005617.3 457 Intron NP_005608.1

View Full Product Details