Product Details

SNP ID
rs1752274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:10423235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCCTGGCCAGAACATACTTATT[T/A]AGCAGGTGTGGATATGTGTATTTAA
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
BAGE2 PubMed Links

Gene Details

Gene
BAGE2
Gene Name
B melanoma antigen family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182482.2 Intron NP_872288.2

View Full Product Details