Product Details

SNP ID
rs7552271
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:28389845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATTGAAAATAGGATTATCATAC[A/G]ATTCAGCAATTCCACTACTGGGTAT
Phenotype
MIM: 608726
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHACTR4 PubMed Links

Gene Details

Gene
PHACTR4
Gene Name
phosphatase and actin regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048183.1 Intron NP_001041648.1
NM_023923.3 Intron NP_076412.3
XM_005245970.2 Intron XP_005246027.1
XM_017002157.1 Intron XP_016857646.1
XM_017002158.1 Intron XP_016857647.1
XM_017002159.1 Intron XP_016857648.1
XM_017002160.1 Intron XP_016857649.1
XM_017002161.1 Intron XP_016857650.1
XM_017002162.1 Intron XP_016857651.1
XM_017002163.1 Intron XP_016857652.1
XM_017002164.1 Intron XP_016857653.1
XM_017002165.1 Intron XP_016857654.1
XM_017002166.1 Intron XP_016857655.1
XM_017002167.1 Intron XP_016857656.1
XM_017002168.1 Intron XP_016857657.1
XM_017002169.1 Intron XP_016857658.1
XM_017002170.1 Intron XP_016857659.1
XM_017002171.1 Intron XP_016857660.1
XM_017002172.1 Intron XP_016857661.1
XM_017002173.1 Intron XP_016857662.1

View Full Product Details