Product Details

SNP ID
rs28610118
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:152511594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCCAAATGTACCCCTAAATG[C/T]CCTCCCAAGTGTACTCCTAAGTGTC
Phenotype
MIM: 612619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
There are no transcripts associated with this gene.

Gene
LCE5A
Gene Name
late cornified envelope 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178438.4 236 Silent Mutation TGC,TGT C20C NP_848525.1

View Full Product Details