Product Details

SNP ID
rs6979659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:740093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACACCCCAGAGCTCCCAGGATC[A/C]GTCTGCGGCTGGCTTCAGCCCCACC
Phenotype
MIM: 614864
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAAF5 PubMed Links

Gene Details

Gene
DNAAF5
Gene Name
dynein axonemal assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017802.3 Intron NP_060272.3

View Full Product Details