Product Details

SNP ID
rs4720773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:740731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCCTCTGCCTCAGGCAGCGTCC[A/G]TATGGCAGCACTCAGAGCCGCACCG
Phenotype
MIM: 614864
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAAF5 PubMed Links

Gene Details

Gene
DNAAF5
Gene Name
dynein axonemal assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017802.3 Intron NP_060272.3

View Full Product Details