Product Details

SNP ID
rs28564742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7569870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTTGCAAAGGTATGGTGATGA[A/G]GGAGGAGGGTATATACTCTAGGAAG
Phenotype
MIM: 615359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927391 PubMed Links

Gene Details

Gene
LOC101927391
Gene Name
uncharacterized LOC101927391
There are no transcripts associated with this gene.

Gene
MIOS
Gene Name
meiosis regulator for oocyte development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019005.3 Intron NP_061878.3
XM_005249780.3 Intron XP_005249837.1
XM_005249781.3 Intron XP_005249838.1
XM_005249782.3 Intron XP_005249839.1
XM_005249784.3 Intron XP_005249841.1
XM_011515432.2 Intron XP_011513734.1
XM_017012361.1 Intron XP_016867850.1
XM_017012362.1 Intron XP_016867851.1

View Full Product Details