Product Details

SNP ID
rs28916296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7640286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGGGTCCCTCCCTCCAGCTAC[A/G]GACTTGGGAGCCCATGATTGCGAAC
Phenotype
MIM: 179837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPA3 PubMed Links

Gene Details

Gene
RPA3
Gene Name
replication protein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002947.4 Intron NP_002938.1
Gene
UMAD1
Gene Name
UBAP1-MVB12-associated (UMA) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302348.1 Intron NP_001289277.1
NM_001302349.1 Intron NP_001289278.1
NM_001302350.1 Intron NP_001289279.1

View Full Product Details