Product Details

SNP ID
rs28541336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140730126 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACAGGTGATGAATCTGCTTTCAC[A/G]AAAAGTATCTTTTCTTAATGATTCC
Phenotype
MIM: 164757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRAF PubMed Links

Gene Details

Gene
BRAF
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004333.4 Intron NP_004324.2
XM_005250045.2 Intron XP_005250102.1
XM_017012558.1 Intron XP_016868047.1
XM_017012559.1 Intron XP_016868048.1

View Full Product Details