Product Details

SNP ID
rs28377711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143136507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGTCAAATTTTTCTATTTTCA[A/G]TCCAAGAATCTGAGACATGTACACT
Phenotype
MIM: 176720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIP PubMed Links

Gene Details

Gene
PIP
Gene Name
prolactin induced protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002652.2 Intron NP_002643.1

View Full Product Details