Product Details

SNP ID
rs28404085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94515443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTGATACGGGTTCAT[C/T]AGATTAGAGCTATTGTTTTCCATCA
Phenotype
MIM: 611686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASD1 PubMed Links

Gene Details

Gene
CASD1
Gene Name
CAS1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022900.4 Intron NP_075051.4
XM_006716092.3 Intron XP_006716155.1
XM_006716093.3 Intron XP_006716156.2
XM_011516495.2 Intron XP_011514797.1
XM_017012546.1 Intron XP_016868035.1
XM_017012547.1 Intron XP_016868036.1

View Full Product Details