Product Details
- SNP ID
-
rs28364018
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:66177635 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGTGCTGGGCTTTGGCCTCAGTGA[G/T]TCGGATGGGCGCTGTCCGCGGTGCT
- Phenotype
-
MIM: 122560
MIM: 606469
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114350622] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRH
- Gene Name
- corticotropin releasing hormone
- Gene
- TRIM55
- Gene Name
- tripartite motif containing 55
There are no transcripts associated with this gene.
View Full Product Details