Product Details

SNP ID
rs11546489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130872597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTCCGCAGGAACGCCGCGATG[A/G]CTGCGCAGGGAGAGCCCCAGGTCCA
Phenotype
MIM: 601179
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAN PubMed Links

Gene Details

Gene
RAN
Gene Name
RAN, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300796.1 262 Intron NP_001287725.1
NM_001300797.1 262 Intron NP_001287726.1
NM_006325.4 262 Missense Mutation ACT,GCT T2A NP_006316.1
XM_017019772.1 262 Missense Mutation ACT,GCT T2A XP_016875261.1
XM_017019773.1 262 Intron XP_016875262.1

View Full Product Details