Product Details

SNP ID
rs28566553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31074586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTGTGCGTGTGTGGGAGCGCG[A/G]GAGCCCCCCGACAGCCACCCCTTGG
Phenotype
MIM: 610561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 814 UTR 5 NP_055514.3
XM_005255710.3 814 UTR 5 XP_005255767.1
XM_005255711.3 814 UTR 5 XP_005255768.1
XM_005255712.3 814 UTR 5 XP_005255769.1
XM_011545990.2 814 Intron XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172668.1 814 Intron NP_001166139.1
NM_001172669.1 814 Intron NP_001166140.1
NM_001172670.1 814 Intron NP_001166141.1
NM_024706.4 814 Intron NP_078982.3

View Full Product Details