Product Details

SNP ID
rs28670302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67846931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGGGGGGAAGGGACAGTCGG[C/T]CGCAGACCGCGCTGGGTTGCCGCTG
Phenotype
MIM: 611510 MIM: 605813 MIM: 609119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPT PubMed Links

Gene Details

Gene
CENPT
Gene Name
centromere protein T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025082.3 200 Intron NP_079358.3
XM_006721277.2 200 Intron XP_006721340.1
XM_011523344.1 200 Intron XP_011521646.1
XM_011523345.1 200 Intron XP_011521647.1
XM_017023712.1 200 Intron XP_016879201.1
XM_017023713.1 200 Intron XP_016879202.1
XM_017023714.1 200 Intron XP_016879203.1
XM_017023715.1 200 Intron XP_016879204.1
XM_017023716.1 200 Intron XP_016879205.1
XM_017023717.1 200 Intron XP_016879206.1
Gene
NUTF2
Gene Name
nuclear transport factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322038.1 200 UTR 5 NP_001308967.1
NM_001322039.1 200 UTR 5 NP_001308968.1
NM_001322040.1 200 Intron NP_001308969.1
NM_001322041.1 200 Intron NP_001308970.1
NM_005796.2 200 UTR 5 NP_005787.1
Gene
THAP11
Gene Name
THAP domain containing 11
There are no transcripts associated with this gene.

View Full Product Details