Product Details

SNP ID
rs28450346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71860772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTATGAGGCCAGTGGCTCCACG[C/G]TCACCACAAGGGGTCAGTTTGAATG
Phenotype
MIM: 614301
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATXN1L PubMed Links

Gene Details

Gene
ATXN1L
Gene Name
ataxin 1 like
There are no transcripts associated with this gene.

Gene
ZNF821
Gene Name
zinc finger protein 821
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201552.1 Intron NP_001188481.1
NM_001201553.1 Intron NP_001188482.1
NM_001201554.1 Intron NP_001188483.1
NM_001201556.1 Intron NP_001188485.1
NM_001318238.1 Intron NP_001305167.1
NM_001318239.1 Intron NP_001305168.1
NM_017530.2 Intron NP_060000.1
XM_005256033.2 Intron XP_005256090.1
XM_006721233.1 Intron XP_006721296.1
XM_011523211.2 Intron XP_011521513.1
XM_011523212.2 Intron XP_011521514.1
XM_011523214.2 Intron XP_011521516.1
XM_011523215.2 Intron XP_011521517.1
XM_011523220.2 Intron XP_011521522.1
XM_017023409.1 Intron XP_016878898.1
XM_017023410.1 Intron XP_016878899.1
XM_017023411.1 Intron XP_016878900.1
XM_017023412.1 Intron XP_016878901.1
XM_017023413.1 Intron XP_016878902.1
XM_017023414.1 Intron XP_016878903.1
XM_017023415.1 Intron XP_016878904.1
XM_017023416.1 Intron XP_016878905.1
XM_017023417.1 Intron XP_016878906.1
XM_017023418.1 Intron XP_016878907.1
XM_017023419.1 Intron XP_016878908.1
XM_017023420.1 Intron XP_016878909.1
XM_017023421.1 Intron XP_016878910.1
XM_017023422.1 Intron XP_016878911.1
XM_017023423.1 Intron XP_016878912.1
XM_017023424.1 Intron XP_016878913.1
XM_017023425.1 Intron XP_016878914.1

View Full Product Details