Product Details
- SNP ID
-
rs28591145
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55765707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCAGGAAATACAGAGAAGGAAA[A/G]GTAAGAAAATTAAAATCACATACAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CES1P1
PubMed Links
Gene Details
- Gene
- CES1P1
- Gene Name
- carboxylesterase 1 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details