Product Details

SNP ID
rs28503425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:660647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTCTTCAGTAACAAGGTGAAC[A/C]CCTTCCAGAGGGCTGAGTAGGTACC
Phenotype
MIM: 607427 MIM: 188350
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ENOSF1 PubMed Links

Gene Details

Gene
ENOSF1
Gene Name
enolase superfamily member 1
There are no transcripts associated with this gene.

Gene
TYMS
Gene Name
thymidylate synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071.2 Intron NP_001062.1
Gene
TYMSOS
Gene Name
TYMS opposite strand
There are no transcripts associated with this gene.

View Full Product Details