Product Details

SNP ID
rs28574941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32093592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTTGTTTTACCTTTCTGCTTA[C/T]TTATTACTTGATTAATACAAGGGTG
Phenotype
MIM: 610432 MIM: 616319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF125 PubMed Links

Gene Details

Gene
RNF125
Gene Name
ring finger protein 125
There are no transcripts associated with this gene.

Gene
RNF138
Gene Name
ring finger protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191324.1 Intron NP_001178253.2
NM_016271.4 Intron NP_057355.2
NM_198128.2 Intron NP_937761.1
XM_005258285.1 Intron XP_005258342.1
XM_005258286.1 Intron XP_005258343.1

View Full Product Details