Product Details

SNP ID
rs28438034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35335434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAACCTATAAACAAGCTCCATT[C/T]TGCCTCACTTTTCCACCAGCTGGCC
Phenotype
MIM: 194534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 3083 Intron NP_001295052.1
NM_006965.3 3083 UTR 3 NP_008896.2
XM_005258341.3 3083 UTR 3 XP_005258398.1

View Full Product Details