Product Details

SNP ID
rs35166625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45003028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTCCACAGTTCCTGCCCACTC[A/G]CTCATGCAGGATGAGGTTGGGAGGA
Phenotype
MIM: 604783 MIM: 604758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1 PubMed Links

Gene Details

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
There are no transcripts associated with this gene.

Gene
RELB
Gene Name
RELB proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006509.3 Intron NP_006500.2
XM_005259127.3 Intron XP_005259184.1
XM_005259128.2 Intron XP_005259185.1

View Full Product Details