Product Details

SNP ID
rs28458769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33088181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTTCAGGATCAGACTGTCAGA[A/G]ATGAAAAAGGAAGGTATAAACGATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH1 PubMed Links

Gene Details

Gene
GPATCH1
Gene Name
G-patch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018025.2 150 Missense Mutation AAT,GAT N41D NP_060495.2
XM_006723255.3 150 Missense Mutation AAT,GAT N41D XP_006723318.1

View Full Product Details