Product Details

SNP ID
rs28624501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19407037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGCCTTTAATTTTCTTTCTTC[A/C]TACCAGTTTACTTCTTTCAGTTTTC
Phenotype
MIM: 614997
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GATAD2A PubMed Links

Gene Details

Gene
GATAD2A
Gene Name
GATA zinc finger domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300946.1 Intron NP_001287875.1
NM_017660.3 Intron NP_060130.3
XM_011528105.1 Intron XP_011526407.1
XM_017026901.1 Intron XP_016882390.1
XM_017026902.1 Intron XP_016882391.1
XM_017026903.1 Intron XP_016882392.1
XM_017026904.1 Intron XP_016882393.1
XM_017026905.1 Intron XP_016882394.1
XM_017026906.1 Intron XP_016882395.1
XM_017026907.1 Intron XP_016882396.1
XM_017026908.1 Intron XP_016882397.1
XM_017026909.1 Intron XP_016882398.1
XM_017026910.1 Intron XP_016882399.1
XM_017026911.1 Intron XP_016882400.1

View Full Product Details